Anti-CACNA1G Mouse Monoclonal Antibody [clone: S178A-9]

Proveedor: OriGene
ORIGTA326469EA 645 EUR
ORIGTA326469
Anti-CACNA1G Mouse Monoclonal Antibody [clone: S178A-9]
Anticuerpos
Type: Primary
Antigen: CACNA1G
Clonality: Monoclonal
Clone: S178A-9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR