Anti-PLK1 Rabbit Polyclonal Antibody [clone: Poly6186]

Proveedor: OriGene
ORIGTA328018EA 454 EUR
ORIGTA328018
Anti-PLK1 Rabbit Polyclonal Antibody [clone: Poly6186]
Type: Primary
Antigen: PLK1
Clonality: Polyclonal
Clone: Poly6186
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR