Anti-MCM7 Rabbit Polyclonal Antibody [clone: Poly6027]

Proveedor: OriGene
ORIGTA328015EA 466 EUR
ORIGTA328015
Anti-MCM7 Rabbit Polyclonal Antibody [clone: Poly6027]
Type: Primary
Antigen: MCM7
Clonality: Polyclonal
Clone: Poly6027
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR