Anti-AKT1 Rabbit Polyclonal Antibody [clone: Poly6034]

Proveedor: OriGene
ORIGTA328005EA 182 EUR
ORIGTA328005
Anti-AKT1 Rabbit Polyclonal Antibody [clone: Poly6034]
Type: Primary
Antigen: AKT1
Clonality: Polyclonal
Clone: Poly6034
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR