Anti-XBP1 Rabbit Polyclonal Antibody [clone: Poly6195]

Proveedor: OriGene
ORIGTA328002EA 466 EUR
ORIGTA328002
Anti-XBP1 Rabbit Polyclonal Antibody [clone: Poly6195]
Type: Primary
Antigen: XBP1
Clonality: Polyclonal
Clone: Poly6195
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR