Anti-ADIPOQ Rabbit Polyclonal Antibody [clone: Poly6215]

Proveedor: OriGene
ORIGTA327999EA 454 EUR
ORIGTA327999
Anti-ADIPOQ Rabbit Polyclonal Antibody [clone: Poly6215]
Type: Primary
Antigen: ADIPOQ
Clonality: Polyclonal
Clone: Poly6215
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR