Anti-CAMP Mouse Monoclonal Antibody [clone: H7]

Proveedor: OriGene
ORIGTA327992EA 525 EUR
ORIGTA327992
Anti-CAMP Mouse Monoclonal Antibody [clone: H7]
Anticuerpos
Type: Primary
Antigen: CAMP
Clonality: Monoclonal
Clone: H7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR