Anti-CETN2 Rabbit Polyclonal Antibody [clone: Poly6288]

Proveedor: OriGene
ORIGTA327991EA 466 EUR
ORIGTA327991
Anti-CETN2 Rabbit Polyclonal Antibody [clone: Poly6288]
Type: Primary
Antigen: CETN2
Clonality: Polyclonal
Clone: Poly6288
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR