Anti-KAT2B Mouse Monoclonal Antibody [clone: 15G10]

Proveedor: OriGene
ORIGTA327982EA 316 EUR
ORIGTA327982
Anti-KAT2B Mouse Monoclonal Antibody [clone: 15G10]
Anticuerpos
Type: Primary
Antigen: KAT2B
Clonality: Monoclonal
Clone: 15G10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR