Anti-HIST3H3 Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA327345EA 466 EUR
ORIGTA327345
Anti-HIST3H3 Rabbit Polyclonal Antibody
Anticuerpos
Recommended Dilutions: Western Blot: 1:500-1:2000; Immunochemistry: 1:50-1:200; Immunofluorescence: 1:50-1:200; Immunoprecipitation: 1:50-1:200 ChImmunoprecipitation: 1:50-1:200

Type: Primary
Antigen: HIST3H3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR