Anti-KPNA5 Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA590118EA 670 EUR
ORIGTA590118
Anti-KPNA5 Rabbit Polyclonal Antibody
Anticuerpos
Recommended Dilutions: Western Blot: 1:500; ELISA: 1:100-1:2000; IHC: 1:300; Immunofluorescence: 1:75

Type: Primary
Antigen: KPNA5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR