Anti-IL15RA Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA590115EA 568 EUR
ORIGTA590115
Anti-IL15RA Rabbit Polyclonal Antibody
Recommended Dilutions: Western Blot: 1:5000-1:20000; ELISA: 1:100-1:2000

Type: Primary
Antigen: IL15RA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR