Anti-AHSA1 Rat Monoclonal Antibody [clone: 25F2.D10]

Proveedor: OriGene
ORIGTA326465EA 713 EUR
ORIGTA326465
Anti-AHSA1 Rat Monoclonal Antibody [clone: 25F2.D10]
Anticuerpos
Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: AHSA1
Clonality: Monoclonal
Clone: 25F2.D10
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a kappa
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR