Anti-Adam22 Mouse Monoclonal Antibody [clone: S57-2]

Proveedor: OriGene
ORIGTA326463EA 574 EUR
ORIGTA326463
Anti-Adam22 Mouse Monoclonal Antibody [clone: S57-2]
Recommended Dilutions: Western Blot: 1?g/ml

Type: Primary
Antigen: Adam22
Clonality: Monoclonal
Clone: S57-2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR