Anti-Abcc9 Mouse Monoclonal Antibody [clone: S319A-14]

Proveedor: OriGene
ORIGTA326461EA 645 EUR
ORIGTA326461
Anti-Abcc9 Mouse Monoclonal Antibody [clone: S319A-14]
Anticuerpos
Type: Primary
Antigen: Abcc9
Clonality: Monoclonal
Clone: S319A-14
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2A
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR