Anti-BBC3 Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA326460EA 645 EUR
ORIGTA326460
Anti-BBC3 Rabbit Polyclonal Antibody
Anticuerpos
Recommended Dilutions: Western Blot: 1:500 Immunochemistry: 1:100

Type: Primary
Antigen: BBC3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR