Anti-APP Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA326459EA 620 EUR
ORIGTA326459
Anti-APP Rabbit Polyclonal Antibody
Anticuerpos
  • Recommended Dilutions: Immunochemistry: 1:1000-1:10000; Immunoprecipitation: 1:1000

Type: Primary
Antigen: APP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: eukaryotes, Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR