Anti-SCN9A Mouse Monoclonal Antibody [clone: S68-6]

Proveedor: OriGene
ORIGTA326452EA 645 EUR
ORIGTA326452
Anti-SCN9A Mouse Monoclonal Antibody [clone: S68-6]
Anticuerpos
Recommended Dilutions: Western Blot: 1-10?g/ml; Immunochemistry: 0.1-1.0?g/ml; Immunofluorescence: 1.0-10?g/ml

Type: Primary
Antigen: SCN9A
Clonality: Monoclonal
Clone: S68-6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR