Anti-HSP90AA1 Mouse Monoclonal Antibody [clone: D7A]

Proveedor: OriGene
ORIGTA326447EA 665 EUR
ORIGTA326447
Anti-HSP90AA1 Mouse Monoclonal Antibody [clone: D7A]
Anticuerpos
Recommended Dilutions: Western Blot: 1:500; Immunoprecipitation: 5?g with 20?l Protein A beads

Type: Primary
Antigen: HSP90AA1
Clonality: Monoclonal
Clone: D7A
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Bovine, Chicken, Human, Mouse, Pig (Pig), Rabbit, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR