Anti-ANK2 Mouse Monoclonal Antibody [clone: S105-13]

Proveedor: OriGene
ORIGTA326443EA 692 EUR
ORIGTA326443
Anti-ANK2 Mouse Monoclonal Antibody [clone: S105-13]
Anticuerpos
Recommended Dilutions: Immunochemistry: 1:50-1:200

Type: Primary
Antigen: ANK2
Clonality: Monoclonal
Clone: S105-13
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR