Anti-ADRA2C Mouse Monoclonal Antibody [clone: S330A-80]

Proveedor: OriGene
ORIGTA326436EA 692 EUR
ORIGTA326436
Anti-ADRA2C Mouse Monoclonal Antibody [clone: S330A-80]
Anticuerpos
Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: ADRA2C
Clonality: Monoclonal
Clone: S330A-80
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR