Anti-ADRA2C Mouse Monoclonal Antibody [clone: S330A-51]

Proveedor: OriGene
ORIGTA326435EA 574 EUR
ORIGTA326435
Anti-ADRA2C Mouse Monoclonal Antibody [clone: S330A-51]
Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: ADRA2C
Clonality: Monoclonal
Clone: S330A-51
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2A
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR