Anti-CRYAB Rabbit Polyclonal Antibody [clone: N/A]

Proveedor: OriGene
ORIGTA326430EA 564 EUR
ORIGTA326430
Anti-CRYAB Rabbit Polyclonal Antibody [clone: N/A]
Anticuerpos
Recommended Dilutions: Western Blot: 1:5000-10000

Type: Primary
Antigen: CRYAB
Clonality: Polyclonal
Clone: N/A
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Chicken, Bovine, Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR