Anti-CRYAB Mouse Monoclonal Antibody [clone: 1A7.D5]

Proveedor: OriGene
ORIGTA326428EA 614 EUR
ORIGTA326428
Anti-CRYAB Mouse Monoclonal Antibody [clone: 1A7.D5]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: CRYAB
Clonality: Monoclonal
Clone: 1A7.D5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Bovine, Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR