Anti-TUBG1 Mouse Monoclonal Antibody [clone: 14C11]

Proveedor: OriGene
ORIGTA327939EA 280 EUR
ORIGTA327939
Anti-TUBG1 Mouse Monoclonal Antibody [clone: 14C11]
Anticuerpos
Type: Primary
Antigen: TUBG1
Clonality: Monoclonal
Clone: 14C11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR