Anti-GFAP Mouse Monoclonal Antibody [clone: 2E1.E9]

Proveedor: OriGene
ORIGTA327938EA 738 EUR
ORIGTA327938
Anti-GFAP Mouse Monoclonal Antibody [clone: 2E1.E9]
Anticuerpos
Type: Primary
Antigen: GFAP
Clonality: Monoclonal
Clone: 2E1.E9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR