Anti-CDK7 Rabbit Polyclonal Antibody [clone: Poly6324]

Proveedor: OriGene
ORIGTA327937EA 466 EUR
ORIGTA327937
Anti-CDK7 Rabbit Polyclonal Antibody [clone: Poly6324]
Type: Primary
Antigen: CDK7
Clonality: Polyclonal
Clone: Poly6324
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR