Anti-APC Rabbit Polyclonal Antibody [clone: Poly6109]

Proveedor: OriGene
ORIGTA327926EA 182 EUR
ORIGTA327926
Anti-APC Rabbit Polyclonal Antibody [clone: Poly6109]
Type: Primary
Antigen: APC
Clonality: Polyclonal
Clone: Poly6109
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR