Anti-CDK4 Rabbit Polyclonal Antibody [clone: Poly6333]

Proveedor: OriGene
ORIGTA327917EA 466 EUR
ORIGTA327917
Anti-CDK4 Rabbit Polyclonal Antibody [clone: Poly6333]
Type: Primary
Antigen: CDK4
Clonality: Polyclonal
Clone: Poly6333
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR