Anti-HSPE1 Rabbit Polyclonal Antibody [clone: N/A]
Proveedor: OriGene
ORIGTA326425EA 534 EUR
ORIGTA326425
Anti-HSPE1 Rabbit Polyclonal Antibody [clone: N/A]
Anticuerpos
Recommended Dilutions: Western Blot: 1:1000
Type: Primary
Antigen: HSPE1
Clonality: Polyclonal
Clone: N/A
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bovine, canine, Guinea Pig, Human, Mouse, Pig, Rabbit, Rat, Sheep, Xenopus