Anti-SOD3 Mouse Monoclonal Antibody [clone: 4GG11G6]

Proveedor: OriGene
ORIGTA326420EA 652 EUR
ORIGTA326420
Anti-SOD3 Mouse Monoclonal Antibody [clone: 4GG11G6]
Anticuerpos
Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:100

Type: Primary
Antigen: SOD3
Clonality: Monoclonal
Clone: 4GG11G6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR