Anti-AARSD1 Mouse Monoclonal Antibody [clone: Tsp232A]

Proveedor: OriGene
ORIGTA326408EA 582 EUR
ORIGTA326408
Anti-AARSD1 Mouse Monoclonal Antibody [clone: Tsp232A]
Anticuerpos
Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: AARSD1
Clonality: Monoclonal
Clone: Tsp232A
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR