Anti-SNAP25 Mouse Monoclonal Antibody [clone: SP-12]

Proveedor: OriGene
ORIGTA326406EA 519 EUR
ORIGTA326406
Anti-SNAP25 Mouse Monoclonal Antibody [clone: SP-12]
Recommended Dilutions: Western Blot: 1:1000 ImmunochemistryC/Immunofluorescence: 1:1000; ELISA: 1:5000

Type: Primary
Antigen: SNAP25
Clonality: Monoclonal
Clone: SP-12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: cat, Human, Mouse, Pig, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR