Anti-AHSA2 Rabbit Polyclonal Antibody [clone: N/A]

Proveedor: OriGene
ORIGTA326404EA 625 EUR
ORIGTA326404
Anti-AHSA2 Rabbit Polyclonal Antibody [clone: N/A]
Anticuerpos
Type: Primary
Antigen: AHSA2
Clonality: Polyclonal
Clone: N/A
Conjugation: Unconjugated
Epitope: Full length
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR