Anti-HSPH1 Rabbit Polyclonal Antibody [clone: N/A]

Proveedor: OriGene
ORIGTA326392EA 554 EUR
ORIGTA326392
Anti-HSPH1 Rabbit Polyclonal Antibody [clone: N/A]
Anticuerpos
Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: HSPH1
Clonality: Polyclonal
Clone: N/A
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bovine, Hamster, Human, Monkey, Mouse, Rat, Sheep, Yeast
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR