Anti-PTGES3 Mouse Monoclonal Antibody [clone: JJ6]

Proveedor: OriGene
ORIGTA326391EA 545 EUR
ORIGTA326391
Anti-PTGES3 Mouse Monoclonal Antibody [clone: JJ6]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: PTGES3
Clonality: Monoclonal
Clone: JJ6
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Chicken, Guinea Pig, Human, Mouse, Rabbit, s. cerevisiae (lower)
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR