Anti-TP53 Mouse Monoclonal Antibody [clone: Pab 1801]

Proveedor: OriGene
ORIGTA326387EA 478 EUR
ORIGTA326387
Anti-TP53 Mouse Monoclonal Antibody [clone: Pab 1801]
Recommended Dilutions: Western Blot: 1:1000-1:2000; Immunoprecipitation: 10?g/mg; Immunochemistry: 1:100-1:250

Type: Primary
Antigen: TP53
Clonality: Monoclonal
Clone: Pab 1801
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR