Anti-SOD2 Rabbit Polyclonal Antibody [clone: N/A]

Proveedor: OriGene
ORIGTA326385EA 699 EUR
ORIGTA326385
Anti-SOD2 Rabbit Polyclonal Antibody [clone: N/A]
Anticuerpos
Recommended Dilutions: 0.2?g/ml

Type: Primary
Antigen: SOD2
Clonality: Polyclonal
Clone: N/A
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bovine, canine, Chicken, gerbil, Guinea Pig, Hamster, Human, Monkey, Mouse, Pig, Rabbit, Rat, Sheep, Xenopus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR