Anti-KCNJ4 Mouse Monoclonal Antibody [clone: S25-35]

Proveedor: OriGene
ORIGTA326378EA 574 EUR
ORIGTA326378
Anti-KCNJ4 Mouse Monoclonal Antibody [clone: S25-35]
Recommended Dilutions: Western Blot: 1-10?g/ml; Immunochemistry: 0.1-1.0?g/ml; Immunofluorescence: 1.0-10?g/ml

Type: Primary
Antigen: KCNJ4
Clonality: Monoclonal
Clone: S25-35
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR