Anti-DNAJB1 Rabbit Polyclonal Antibody [clone: N/A]

Proveedor: OriGene
ORIGTA326377EA 682 EUR
ORIGTA326377
Anti-DNAJB1 Rabbit Polyclonal Antibody [clone: N/A]
Anticuerpos
Recommended Dilutions: Western Blot: 1:1500-2000

Type: Primary
Antigen: DNAJB1
Clonality: Polyclonal
Clone: N/A
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: African Green Monkey, Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR