Anti-DNAJB1 Mouse Monoclonal Antibody [clone: 3B9.E6]

Proveedor: OriGene
ORIGTA326376EA 627 EUR
ORIGTA326376
Anti-DNAJB1 Mouse Monoclonal Antibody [clone: 3B9.E6]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: DNAJB1
Clonality: Monoclonal
Clone: 3B9.E6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR