Anti-IRS1 Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA327864EA 506 EUR
ORIGTA327864
Anti-IRS1 Rabbit Polyclonal Antibody
Anticuerpos
Recommended Dilutions: Immunochemistry: 1:50-1:200

Type: Primary
Antigen: IRS1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR