Anti-HIST1H2AM Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA327846EA 525 EUR
ORIGTA327846
Anti-HIST1H2AM Rabbit Polyclonal Antibody
Anticuerpos
Recommended Dilutions: Immunochemistry: 1:50-1:200

Type: Primary
Antigen: HIST1H2AM
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR