Anti-ESR2 Rabbit Polyclonal Antibody

Proveedor: OriGene
ORIGTA327832EA 564 EUR
ORIGTA327832
Anti-ESR2 Rabbit Polyclonal Antibody
Anticuerpos
Recommended Dilutions: Western Blot: 1:500-1:2000; Immunofluorescence/Immunochemistry: 1:100-1:500

Type: Primary
Antigen: ESR2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR