Anti-HSP90AB1 Rabbit Polyclonal Antibody [clone: N/A]
Proveedor: OriGene
ORIGTA326372EA 596 EUR
ORIGTA326372
Anti-HSP90AB1 Rabbit Polyclonal Antibody [clone: N/A]
Anticuerpos
Recommended Dilutions: Western Blot: 1:20000-40000
Type: Primary
Antigen: HSP90AB1
Clonality: Polyclonal
Clone: N/A
Conjugation: Unconjugated
Epitope: Full length
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat