Anti-HSP90AA1 Mouse Monoclonal Antibody [clone: 2G5.G3]

Proveedor: OriGene
ORIGTA326368EA 713 EUR
ORIGTA326368
Anti-HSP90AA1 Mouse Monoclonal Antibody [clone: 2G5.G3]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: HSP90AA1
Clonality: Monoclonal
Clone: 2G5.G3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Mouse, Rat.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR