Anti-HSPB1 Mouse Monoclonal Antibody [clone: 5D12-A12]

Proveedor: OriGene
ORIGTA326365EA 582 EUR
ORIGTA326365
Anti-HSPB1 Mouse Monoclonal Antibody [clone: 5D12-A12]
Anticuerpos
Recommended Dilutions: Western Blot: 0.25-0.5 ?g/ml

Type: Primary
Antigen: HSPB1
Clonality: Monoclonal
Clone: 5D12-A12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR