Anti-HSPA1A Rabbit Polyclonal Antibody [clone: N/A]

Proveedor: OriGene
ORIGTA326361EA 682 EUR
ORIGTA326361
Anti-HSPA1A Rabbit Polyclonal Antibody [clone: N/A]
Anticuerpos
Recommended Dilutions: Western Blot: 1:25000; Immunoprecipitation: 1:100

Type: Primary
Antigen: HSPA1A
Clonality: Polyclonal
Clone: N/A
Conjugation: Unconjugated
Epitope: Full length
Host: Rabbit
Isotype:
Reactivity: atlantic hagFish (myxine glutinosa), beluga, coral, Bovine, Dog, Fish (carp), Guinea Pig, Hamster, Human, Monkey, Mouse, Pig, Rat, Sheep, spiny DogFish shark (squalus acanthias), tobacco, tomato
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR