Anti-EPAS1 Mouse Monoclonal Antibody [clone: EP190B]

Proveedor: OriGene
ORIGTA326351EA 519 EUR
ORIGTA326351
Anti-EPAS1 Mouse Monoclonal Antibody [clone: EP190B]
Recommended Dilutions: Western Blot: 1:100-1:500

Type: Primary
Antigen: EPAS1
Clonality: Monoclonal
Clone: EP190B
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR