Anti-MAPK14 Mouse Monoclonal Antibody [clone: 9F12]

Proveedor: OriGene
ORIGTA326349EA 577 EUR
ORIGTA326349
Anti-MAPK14 Mouse Monoclonal Antibody [clone: 9F12]
Anticuerpos
Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: MAPK14
Clonality: Monoclonal
Clone: 9F12
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR